missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxybenzenesulfonic acid, 65%

Product Code. 15406368
Change view
Click to view available options
Quantity:
250 mL
1 L
Unit Size:
1 L
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15406368 250 mL 250mL
15416368 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15406368 Supplier Thermo Scientific Alfa Aesar Supplier No. H59917.AK
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 98-67-9 | C6H6O4S | 174.17 g/mol

4-Hydroxybenzenesulfonic Acid is used as a redox-mediator in the laccase-catalyzed degradation of indigo, a textile dye. Also used in oilfield chemicals, metal working, and as a polymerization catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxybenzenesulfonic Acid is used as a redox-mediator in the laccase-catalyzed degradation of indigo, a textile dye. Also used in oilfield chemicals, metal working, and as a polymerization catalyst.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-67-9
Molecular Formula C6H6O4S
Molecular Weight (g/mol) 174.17
MDL Number MFCD00007506
InChI Key FEPBITJSIHRMRT-UHFFFAOYSA-N
Synonym p-phenolsulfonic acid, phenolsulfonic acid, 4-phenolsulfonic acid, p-sulfophenol, sulfocarbolic acid, benzenesulfonic acid, 4-hydroxy, p-hydroxybenzenesulfonic acid, phenol-4-sulfonic acid, benzenesulfonic acid, p-hydroxy, 4-hydroxyphenylsulfonic acid
PubChem CID 4765
ChEBI CHEBI:32354
IUPAC Name 4-hydroxybenzenesulfonic acid
SMILES OC1=CC=C(C=C1)S(O)(=O)=O

Specifications

Density 1.337 g/mL
Refractive Index 1.489
Quantity 250 mL
UN Number UN1803
Beilstein 1869034
Solubility Information Soluble in water.
Formula Weight 174.17
Percent Purity 65%
Physical Form Liquid
Chemical Name or Material 4-Hydroxybenzenesulfonic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.