accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 99-91-2

H3COCl

CAS RN 99-91-2

IUPAC Name: 1-(4-chlorophenyl)ethan-1-one
Synonyms: 4-chloroacetophenone C8H7ClO usaf DO-1
Molecular Weight (g/mol): 154.59
Molecular Formula: C8H7ClO
InChi Key: BUZYGTVTZYSBCU-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(Cl)C=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)