Outcome Certainty Chemicals
Ensuring Measurement Precision: pH Buffers and Conductivity Standards
November 2026 | 8 min read | By: Mettler Toledo
Ensuring Measurement Precision: pH Buffers and Conductivity Standards
We often hear about the importance of accurate pH and conductivity measurements for ensuring the reliability and quality of various analytical processes. Ever wonder what makes those readings reliable? Central to achieving this accuracy is the proper calibration of pH and conductivity sensors using reference solutions known as buffers and conductivity standards which have precisely defined values. These calibration solutions, offered by numerous producers, come with specified quality levels that determine their measurement uncertainty.
Given that typical pH measurement errors range around ±0.05 pH units, any deviation from the stated uncertainty of these calibration buffers introduce additional, often unnoticed, errors into every pH measurement performed. This is why it is essential to use high-quality buffers that are traceable back to certified reference materials to maintain measurement integrity.
So, what exactly are these calibration solutions, and why do they matter so much?
pH Buffers
pH buffers are aqueous solutions that resist variations in pH levels when small amounts of acid or base are added. Every pH level corresponds to a specific buffer capacity and buffer range. This means that at a given pH, a buffer solution has a defined ability to resist changes in pH when acids or bases are added (buffer capacity), and it effectively maintains its pH within a certain range around that value (buffer range).
Buffer capacity describes how much acid or base can be added to the buffer before the pH starts to change noticeably. The larger the buffer capacity, the more resistant the solution is to changes in pH when substances are added.
Buffer range is the window of pH values where the buffer solution can work efficiently—outside this range, the buffer quickly loses its ability to resist pH changes.
Conductivity Standards
A conductivity standard is a reference solution of a known conductivity value that is used for sensor calibration or verification. Using the appropriate conductivity standard ensures accurate measurements. For calibration, higher conductivity standards are typically used to determine the cell constant of the sensor, while the low conductivity standards, which require special handling, are mainly used for verification purposes.
Where are calibration solutions used? 
Calibration solutions are commonly used in various scientific and industrial applications for calibrating pH or conductivity sensors. At METTLER TOLEDO, pH buffers are available in two packaging formats: single-use buffer sachets or bottles of buffer solution. Some of the common applications are as follows:
Laboratory Research
Calibration solutions are essential in laboratory settings to ensure the accuracy and reliability of not only pH meters but also measuring instruments such as conductivity meters. By providing known reference values, these solutions help verify and adjust the instruments to produce precise measurements during experiments and analyses.
Quality Control in Manufacturing
In industrial manufacturing processes, calibration solutions are used regularly to maintain instrument performance, ensuring consistent product quality. Regular calibration helps detect instrument drift or malfunctions early, minimizing errors in measurements critical for production control.
Environmental Monitoring
Environmental laboratories use calibration solutions to confirm the precision of instruments measuring water quality, soil properties, and air samples.  Accurate readings are vital for compliance with environmental regulations and assessing pollution levels.
Pharmaceutical
Buffers and solutions  are crucial for maintaining the stability and efficacy of drugs during formulation and testing. Buffers help maintain the desired pH in drug formulations, ensuring the active ingredients remain stable and effective. 
Food and Beverage Industry
In food processing, calibration solutions are used to ensure instruments measuring acidity, salinity, or other chemical properties deliver accurate results. This helps maintain product consistency, safety, and compliance with health standards.
What does METTLER TOLEDO offer when it comes to pH buffer solutions? 
Be it technical buffers, certified buffers or NIST/DIN buffers, METTLER TOLEDO offers a complete portfolio of pH calibration solutions for any application.
Technical buffers come with a certificate from METTLER TOLEDO with a precision of ±0.02 pH units for most solutions.
Product CodeSupplier No.DescriptionUnit Size
10598281
51350002Technical Buffer pH 2.00250 mL
1260130651350016Technical Buffer pH 2.006x 250 mL
1533940030111134Technical Buffer pH 2.0030x 20 mL (sachets)
1020184151350004Technical Buffer pH 4.01250 mL
1261130651350018Technical Buffer pH 4.016x 250 mL
1266351651302069Technical Buffer pH 4.0130x 20 mL (sachets)
1747555930464188Technical Buffer pH 5.00250 mL
1068370151350006Technical Buffer pH 7.00250 mL
1262130651350020Technical Buffer pH 7.006x 250 mL
1272316151302047Technical Buffer pH 7.0030x 20 mL (sachets)
1722497330464189Technical Buffer pH 8.00250 mL
1038319151350008Technical Buffer pH 9.21250 mL
1263130651350022Technical Buffer pH 9.216x 250 mL
1213780051302070Technical Buffer pH 9.2130x 20 mL (sachets)
1034329151350010Technical Buffer pH 10.00250 mL
1042270151350024Technical Buffer pH 10.006x 250 mL
1043172151302079Technical Buffer pH 10.0130x 20 mL (sachets)
1063409151350012Technical Buffer pH 11.00250 mL
1196253551350026Technical Buffer pH 11.006x 250 mL
1539569830111135Technical Buffer pH 11.0030x 20 mL (sachets)
1059757151302068Rainbow sachets I (4.01 / 7.00 / 9.21)3x 10x 20 mL (sachets)
1269351651302080Rainbow sachets II (4.01 / 7.00 / 10.01)3x 10x 20 mL (sachets)
1536103430095312Rainbow bottles I (4.01 / 7.00 / 9.21)3x 2x 250 mL
Certified buffers are verified and certified by DAkkS (Deutsche Akkreditierungsstelle, German Accreditation Body) accredited laboratory, with a precision of ±0.02 pH units. These buffers are traceable to a certificate of an accredited body of DKD (Deutscher Kalibrierdienst, German Calibration Service). These are measured according to ISO/IEC 17025. 
Product CodeSupplier No.DescriptionUnit size
1023842251350032Certified Buffer pH 4.01250 mL
1071176551350042Certified Buffer pH 4.016x 250 mL
1070988351350034Certified Buffer pH 7.00250 mL
1071195551350044Certified Buffer pH 7.006x 250 mL
1053335351350036Certified Buffer pH 9.21250 mL
1065724351350046Certified Buffer pH 9.216x 250 mL
1068486351350038Certified Buffer pH 10.00250 mL
1265130651350048Certified Buffer pH 10.006x 250 mL
NIST/DIN buffers are prepared according to DIN/ISO 19266 and follow the recommendation of the National Institute of Standards and Technology and the German Institute of Standardization, with a precision of ±0.015 pH units.
Product CodeSupplier No.DescriptionUnit Size
1717568130458274NIST/DIN Buffer pH 1.679250 mL
1755578630667828NIST/DIN Buffer pH 1.67930x 20 mL (sachets)
1269923551350052NIST/DIN Buffer pH 4.006250 mL
1539226530111136NIST/DIN Buffer pH 4.00630x 20 mL (sachets)
1047407151350054NIST/DIN Buffer pH 6.865250 mL
1531227530111137NIST/DIN Buffer pH 6.86530x 20 mL (sachets)
1261924551350056NIST/DIN Buffer pH 9.180250 mL
1532227530111138NIST/DIN Buffer pH 9.18030x 20 mL (sachets)
1064313151350058NIST/DIN Buffer pH 10.012250 mL
1780377530111139NIST/DIN Buffer pH 10.01230x 20 mL (sachets)
1740663330464127NIST/DIN Buffer pH 12.454250 mL
Conductivity standards including 5 µS/cm, 10 µS/cm, 84 µS/cm, 500 µS/cm, 1413 µS/cm, and 12.88 mS/cm can help with the calibration and verification of conductivity sensors. High conductivity standards are the ones with high ion concentration and the low conductivity standards are the ones having low concentration of ions. However, these standards tend to be affected by dilution, contamination, or the influence of CO2 when in contact with air; their values change readily, especially in the case of low conductivity standards. This is why a conductivity standard cannot be stored or used for an extended period after opening. 
The 10 µS/cm, 1.3 µS/cm, 5 µS/cm standards are intended only for verification of a measurement near the pure water range as the values of lower conductivity standards are easily affected by any mishandling. On the other hand, the 84 µS/cm standard should be used for the best calibration accuracy. 
Product CodeSupplier No.DescriptionUnit Size
1545303330090847Conductivity Check Solution 1.3 µS250 mL
1589965730094617Conductivity Standard 5 µS250 mL
1054528451300169Conductivity Standard 10 µS250 mL
1195253551302153Conductivity Standard 84 µS250 mL
1197750151300170Conductivity Standard 500 µS250 mL
1056376251350092Conductivity Standard 1413 µS250 mL
1260131651350096Conductivity Standard 1413 µS6x 250 mL
1016638051302049Conductivity Standard 1413 µS30x 20 mL (sachets)
1025935151350094Conductivity Standard 12.88 mS250 mL
1261131651350098Conductivity Standard 12.88 mS6x 250 mL
1052863251302050Conductivity Standard 12.88 mS30x 20 mL (sachets)