Cold Resistant Gloves
Cold-Resistant Gloves
Gloves designed to protect against a range of cold temperatures while working outside, or with freezer/refrigeration units or cryogenic storage (suitable for handling dry ice, liquid nitrogen, or liquid oxygen). Regulatory compliance details can be obtained at the individual product level.
FEATURED
Thermo Scientific™ Cryo Gloves - Mid-Arm Length
Safely handle samples from ultra-low and cryogenic storage with these cryo gloves which offer a high level of thermal protection.
      Thermo Scientific™ Cryo Gloves - Mid-Arm Length
      FIGHTS FROSTBITE
      Ansell™ ActivArmr™ 97-631
      Feel everything but the cold
      Industry leading combination of comfort, warmth, and dexterity. Food approved.
      Ansell™ ActivArmr™ 97-631
      SPLASH PROTECTION
      Thermo Scientific™ Waterproof Cryo Gloves - Mid-Arm Length
      Comfortable, lightweight, and breathable with splash protection
      Safely handle samples from ultra low temperature freezers and cryogenic storage.
      Thermo Scientific™ Waterproof Cryo Gloves - Mid-Arm Length
      Complete Your Order - Great Deals
      FAQ
      The best material for cold-resistant gloves depends on the specific requirements, including the level of cold, the environment, and the activities you'll be performing. Here are some commonly used materials for cold-resistant gloves and their key properties:
      • Insulating Liners (Thinsulate, Fleece, Wool)
        • Thinsulate is known for its high warmth-to-thickness ratio, making gloves less bulky while providing excellent insulation.
        • Fleece is soft, comfortable, and provides good insulation and moisture-wicking properties.
        • Wool is a natural fiber that offers excellent insulation, even when wet, and is breathable.
      • Leather (Cowhide, Deerskin, Goatskin)
      Durable, provides good abrasion resistance, and offers moderate thermal protection. Often used with an insulating liner for added warmth. Best for general outdoor work, construction, and activities requiring durability.
      • Synthetic Leather 
      Offers similar benefits to natural leather but can be more flexible and durable in wet conditions. Best for tasks requiring dexterity and durability in wet or moderately cold conditions.
      • Neoprene 
      Excellent insulation, waterproof, flexible, and resistant to chemicals. Provides good protection in wet and cold environments. Best for fishing, handling wet materials, or working in cold, damp conditions.
      • Nitrile-Coated Gloves
      Provides a waterproof barrier with good flexibility and moderate thermal protection when combined with an insulating liner. Best for tasks requiring dexterity in cold and wet conditions.
      • PVC (Polyvinyl Chloride)
      Waterproof, provides good insulation and flexibility. Suitable for wet and extremely cold environments. Best for handling ice and snow, or working in wet, cold conditions.
      For temperatures as extreme as -196°C, such as when handling liquid nitrogen, specialized cryogenic gloves are required. These gloves are designed to provide protection against extremely low temperatures and prevent frostbite or cold burns. Here are the key features and materials to look for in cryogenic gloves suitable for such conditions:
      • Multiple Layers of Insulation
      Materials often include layers of neoprene, Thinsulate, or other advanced insulating materials. Multiple layers trap air and provide superior thermal insulation.
      • Waterproof and Chemical-Resistant Outer Layer
      Typically made from materials like neoprene or specially treated fabrics. Protects against liquid nitrogen spills and other cryogenic fluids.
      • Flexible and Dexterous
      Despite the multiple layers, the gloves should allow for some degree of flexibility and dexterity to handle tools and materials safely.
      • Extended Cuffs
      Longer cuffs provide additional protection for the wrists and lower arms, preventing exposure to cryogenic liquids.
      • Certification and Standards
      Ensure the gloves meet relevant safety standards and certifications for cryogenic protection, such as those set by ASTM (American Society for Testing and Materials) or EN (European Norms).
      When selecting cold-resistant gloves, it's important to consider several factors to ensure they meet your specific needs and provide adequate protection. Here are five key things to consider:
      1. Temperature Range
      Ensure the gloves are rated for the specific temperature range you will be exposed to. Look for gloves with appropriate insulation materials and thermal ratings suitable for the coldest conditions you expect to encounter.
      2. Waterproofing
      Protects your hands from moisture, which can quickly lead to cold-related injuries. Choose gloves made from waterproof materials or those with waterproof membranes if you will be working in wet or snowy conditions.
      3. Dexterity and Fit
      Ensures you can perform tasks efficiently without removing the gloves. Look for gloves that offer a good balance between warmth and dexterity. Ensure they fit well, allowing for comfortable movement and grip.
      4. Durability and Abrasion Resistance
      Extends the lifespan of the gloves and protects your hands from mechanical hazards. Choose gloves made from durable materials like leather, synthetic leather, or reinforced fabrics if you will be handling rough or abrasive materials.
      5. Specific Use Case
      Different activities and environments require different types of gloves. Select gloves designed for your specific activity, whether it's industrial work, outdoor sports, or handling cryogenic materials. Specialized gloves will offer features tailored to those tasks, such as enhanced grip, impact protection, or chemical resistance.
      Resources and Related Information
      Safety First... and Fast
      Safety starts here - find information on hand, eye, face, and hearing protection items, as well as a range of other safety items.
      Safety First... and Fast
      Indefinite Sample Storage. Infinite Possibilities.
      Thermo Scientific™ solutions for cryopreservation sample preparation and storage.
      Indefinite Sample Storage. Infinite Possibilities.
      PPE Selection, Fit, and Maintenance
      Find information on product selection, fit, usage, and maintenance for PPE items.
      PPE Selection, Fit and Maintenance Tools
      Shop By Brand