General Chemistry Solutions
General Chemistry Solutions
General Chemistry Solutions help laboratories keep results consistent, comparable, and audit-ready. From NIST-traceable titration standards to ready-to-use acids, bases, oxidizers, and buffers, these solutions reduce preparation time while supporting dependable concentration and pH control. Explore a broad selection for routine QC, teaching labs, and research workflows—ideal for volumetric analysis, calibration, and method verification. Shop trusted suppliers including Thermo Scientific Acros, Fisher Chemicals, Chem-Lab, Thermo Scientific Alfa Aesar, Reagecon, and Fisher Bioreagents to find the right specification for your application, with clear product details and direct ordering.
Silver Nitrate Solution, 0.1M (0.1N), NIST Standard Solution Fisher Chemical
Ready to use, for volumetric analysis, meets analytical specification of Ph.Eur., BP.
Silver Nitrate Solution 0.1M (0.1N), NIST Standard Solution Fisher Chemical
Hydrogen Chloride, AcroSeal™
4N Solution in 1,4-dioxane.
Hydrogen chloride, AcroSeal™
Hydrogen Peroxide 6%, Fisher Chemical™
Hydrogen peroxide 6% (w/v) (20 volumes), extra pure SLR.
Hydrogen Peroxide 6%, Fisher Chemical™
Complete Your Order - Great Deals
Ready-to-use Solutions for Routine Analysis
Keep workflows moving with consistent concentrations of hydrogen peroxide for prep, calibration, and day-to-day chemistry.
Ready-to-use solutions for routine analysis
Faster QC Starts Here
Reduce in-lab prep time by choosing dependable, ready-to-use solutions of hydrogen peroxide for common methods and checks.
Faster QC starts here
FAQ
General chemistry solutions are used for titration, calibration, and routine analysis where consistent concentration matters. Ready-to-use standards help reduce preparation time and variability. Labs commonly use them for QC checks, method verification, and teaching experiments, often selecting trusted suppliers like Fisher Chemicals, Reagecon, and Thermo Scientific Acros.
Choose the concentration that matches your method and expected sample range to keep titration volumes practical and endpoints clear. For regulated or validated work, select NIST-traceable or certified standards where available. Brands like Fisher Chemicals and Reagecon offer ready-to-use options that support repeatable results and easier documentation.
For calibration and routine QC, ready-to-use NIST-traceable standards and stable buffer solutions are common choices because they reduce operator variation. Look for clear specification, expiry, and storage guidance. Fisher Chemicals and Reagecon are frequently used for volumetric standards, while Thermo Scientific Acros supports specialized applications.
Store solutions according to the SDS and label requirements, typically in tightly closed containers, away from heat and incompatible chemicals. Oxidizers like hydrogen peroxide should be segregated, while acids and bases may require corrosion-resistant storage. Using ready-to-use solutions from Fisher Chemicals or Reagecon helps ensure consistent packaging and documented handling guidance.
Traceability links a solution’s stated concentration to recognized standards, supporting confidence in results and simplifying audits. It’s especially important for regulated labs and inter-lab comparability. NIST-traceable solutions and certified standards—often available from Fisher Chemicals and Reagecon—help reduce measurement uncertainty and improve reproducibility.
Ready-to-use solutions reduce steps where error can occur—such as weighing, dilution, and standardization—so day-to-day results are more consistent. They also help standardize procedures across teams and sites. Selecting quality-controlled solutions from suppliers like Fisher Chemicals, Thermo Scientific Acros, and Reagecon supports reliable performance and faster turnaround.
Top Brands