Water Baths
General Purpose Water Baths
General purpose water baths are a dependable way to heat samples gently and evenly for routine laboratory workflows such as incubation, thawing, reagent warming, and temperature-controlled reactions. Choose from unstirred, stirred, and circulating models to match your required stability, uniformity, and throughput. Explore trusted options from Grant Instruments, Thermo Scientific, Fisherbrand, Lauda, and Memmert to support consistent, repeatable results.
Grant Instruments™ JB Nova Digital Water Bath with Lid
Provides stable temperature control, simple interface and fast heat-up.
Water Baths
Thermo Scientific™ Precision™ General Purpose Baths
Support a wide range of applications with general purpose water baths, designed to maintain temperature from ambient to 100°C.
Water Baths
Fisherbrand™ Isotemp™ General Purpose Deluxe Water Baths
Choose general purpose water baths, designed to provide consistent temperature for reliable results with easy-to-use operation.
Water Baths
Selector Guide
Get the best-fit general purpose water bath for your lab protocols with our interactive selection guide:
Save Now - Exclusive Deals
FAQ
A general purpose water bath provides controlled, gentle heating for routine lab tasks like warming reagents, thawing samples, incubation, and temperature conditioning. Choose the capacity and control type that fits your workflow, with popular options from Grant Instruments, Thermo Scientific, Fisherbrand, Memmert, and Lauda.
Start with your largest common vessel size and the number of samples you run at once. A 5 to 12L unit suits small routine work, while 18 to 28L supports higher throughput. Compare internal dimensions, lid options, and temperature range across brands like Grant Instruments and Thermo Scientific.
Circulating (stirred) baths offer better temperature uniformity across the tank, which helps with sensitive protocols and repeatability. Unstirred baths are simpler and often sufficient for basic warming. Thermo Scientific circulating models and Grant Instruments baths are commonly chosen for consistent results.
Use a fitted lid whenever possible and keep the bath at the lowest effective setpoint. Lids help reduce heat loss and evaporation, improving stability and reducing top-ups. Many Grant Instruments and Fisherbrand models include compatible lids for day-to-day lab convenience.
For routine warming and incubation, consistent stability and easy setpoint control matter more than extreme precision. Digital controllers can improve repeatability and ease of use, especially when setpoints change frequently. Consider digital options from Memmert, Fisherbrand, and Thermo Scientific for reliable daily performance.
Position the water bath on a stable, level surface with good ventilation and easy access for filling and draining. Keep it away from sensitive instruments and ensure safe cable routing. Follow the manufacturer’s guidance—Grant Instruments, Memmert, and Thermo Scientific provide clear safety and setup instructions.
Top Brands