Storage Microplates
Storage Microplates
Storage microplates play an essential role in sample preservation, high-density archiving and automated handling workflows. Designed in SBS-compatible formats, these plates offer reliable sealing surfaces, consistent well geometry and strong temperature tolerance for long-term storage, mixing and compound management.
Thermo Scientific™ Abgene™ 96 Well 0.8mL Polypropylene DeepWell™ Sample Processing and Storage Plate for Genomics and NGS library preparation
Specifically designed for use with liquid handlers, and magnetic bead extraction
Thermo Scientific™ Abgene™ 96 Well 0.8mL Polypropylene DeepWell™ Sample Processing & Storage Plate for Genomics and NGS library preparation
Thermo Scientific™ Abgene™ 96 Well 1.2mL Polypropylene DeepWell™ Sample Processing and Storage Plate
Optimize suspension cell culture with these U-bottomed 1.2 mL storage plates.
Thermo Scientific™ Abgene™ 96 Well 1.2mL Polypropylene DeepWell™ Sample Processing & Storage Plate
Thermo Scientific™ Abgene™ 96 Well 0.8mL Polypropylene DeepWell™ Sample Processing and Storage Plate for Genomics and NGS library preparation
Thermo Scientific™ Abgene™ polypropylene microplates and DeepWell plates offer storage security for assays, compound libraries or storing samples for either intermediate or long term use and have molded slots and pads for automated sample processing and preparation workflows. 
Thermo Scientific™ Abgene™ 96 Well 0.8mL Polypropylene DeepWell™ Sample Processing & Storage Plate for Genomics and NGS library preparation
FAQ
Storage microplates provide secure preservation of liquids, pellets or compounds in SBS-compliant formats suitable for manual or automated workflows. Brands such as Thermo Scientific Nunc, Corning, Axygen, Fisherbrand, Eppendorf and Brand support routine storage, screening and biobanking applications.
Consider sample volume, available freezer space and workflow needs. Deepwell plates suit larger volumes or pellet collection, while low-profile plates optimise storage density. Choose the well format that aligns with mixing, freezing and downstream handling requirements.
Polypropylene is commonly used for its chemical resistance and suitability for freezing and storage. Polystyrene may be chosen when clarity is required. Thermo Scientific Nunc, Corning and Axygen frequently use polypropylene for robust sample preservation.
Most storage microplates follow SBS standards, enabling compatibility with liquid handlers, stackers, readers and sealing systems. Rigid construction and reliable well geometry minimise errors during automated transfers.
Proper sealing prevents evaporation, contamination and sample loss during storage and transport. Using compatible films, heat seals or lids ensures sample integrity, especially for long-term or low-temperature workflows.
Top Brands