missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD5/CD20 Antibody Cocktail, FITC, PE
GREENER_CHOICE

Product Code. 11583603
Change view
Click to view available options
Quantity:
100 Tests
Unit Size:
100 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11583603 100 Tests 100 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11583603 Supplier Invitrogen™ Supplier No. MA112138
  • ONLINE EXCLUSIVE PRICE
    566.00€
    441.48€ / 100 tests
    Save 124.52€
    22% Off
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Antibody Cocktail

This antibody is a monoclonal mixture of FITC-conjugated CD5 (clone M28623, isotype IgG2a) and PE-conjugated CD20 (clone DFA-7, isotype IgG1).

In humans, CD5 is a 67kDa T lymphocyte single chain transmembrane glycoprotein. It is present on all mature T lymphocytes, on most of thymocytes and on many T cell leukemias and lymphomas. It reacts with a subpopulation of activated B cells. The CD20 antigen is present on human pre-B lymphocytes and on B-lymphocytes atall stage of maturation, except on plasma cells. Low level expression of the CD20 antigen has been detected on normal T lymphocytes. CD20 is a non glycosylated protein with a molecular weight of 35 or 37 kDa depending on the degree of phosphorylation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD5/CD20
Applications Flow Cytometry
Classification Cocktail
Clone M28623, DFA-7, DFA-7
Conjugate FITC/PE
Formulation PBS with 0.2% carrier protein and 0.08% sodium azide
Gene MS4A1
Gene Accession No. P06127, P11836
Gene Alias B1; B-lymphocyte antigen CD20; B-lymphocyte cell-surface antigen B1; B-lymphocyte surface antigen B1; Bp35; CD20; CD20 antigen; CD20 receptor; CD5; CD5 antigen (p56-62); CD5 antigen p56-62; CD5 molecule; CVID5; LEU1; LEU-16; Leukocyte surface antigen Leu-16; lymphocyte antigen T1/Leu-1; membrane spanning 4-domains A1; membrane-spanning 4-domains subfamily A member 1; membrane-spanning 4-domains, subfamily A, member 1; MGC3969; MS4A1; MS4A2; S7; T1; T-cell surface glycoprotein CD5
Gene Symbols CD5, MS4A1
Host Species Mouse
Immunogen human T-acute lymphoblastic leukemia (ALL) cells and LB lymphoblastoid cell line.
Purification Method Protein A/G
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 921, 931
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1, IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.