Learn More
1,2,3-Tri-O-acetyl-5-deoxy-beta-D-ribofuranose, 98%
Product Code. 15458329
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15458329 1 g 1g
15468329 5 g 5g
15420657 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15458329 Supplier Thermo Scientific Alfa Aesar Supplier No. J66818.03
Request Bulk or Custom Format
CAS: 62211-93-2 | C11H16O7 | 260.24 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 62211-93-2
Molecular Formula C11H16O7
Molecular Weight (g/mol) 260.24
MDL Number MFCD08458459,MFCD08458459
InChI Key NXEJETQVUQAKTO-UHFFFAOYNA-N
Synonym 1,2,3-triacety1-5-deoxy-beta-d-ribofuranose
PubChem CID 70847275
IUPAC Name [(2R,5S)-4,5-diacetyloxy-2-methyloxolan-3-yl] acetate
SMILES CC1OC(OC(C)=O)C(OC(C)=O)C1OC(C)=O
Specifications
Melting Point 65°C to 69°C
Color Yellow to Brown
Quantity 1 g
Solubility Information Soluble in ethanol
Formula Weight 260.24
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 1,2,3-Tri-O-acetyl-5-deoxy-beta-D-ribofuranose