missing translation for 'onlineSavingsMsg'
Learn More

1,2,4-Benzenetricarboxylic anhydride, 97%

Product Code. 10725171 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
250 g
1 kg
Packaging:
Plastic bottle
Unit Size:
1kg
250g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10725171 25 g Plastic bottle 25g
10497280 250 g Plastic bottle 250g
10395970 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10725171 Supplier Thermo Scientific Acros Supplier No. 105370250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 552-30-7 | C9H4O5 | 192.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 552-30-7
MDL Number MFCD00005925
InChI Key SRPWOOOHEPICQU-UHFFFAOYSA-N
Synonym trimellitic anhydride, 1,2,4-benzenetricarboxylic anhydride, anhydrotrimellic acid, trimellitic acid anhydride, 4-carboxyphthalic anhydride, tman, anhydrotrimellitic acid, trimellic acid anhydride, 1,3-dioxo-1,3-dihydroisobenzofuran-5-carboxylic acid, trimellitic acid 1,2-anhydride
PubChem CID 11089
ChEBI CHEBI:53050
IUPAC Name 1,3-dioxo-2-benzofuran-5-carboxylic acid
SMILES C1=CC2=C(C=C1C(=O)O)C(=O)OC2=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 164°C to 169°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Plastic bottle
Molecular Formula C9H4O5
Quantity 25 g
Beilstein 18, 468
Merck Index 15, 9872
Molecular Weight (g/mol) 192.126
Formula Weight 192.13
Percent Purity ≥96.0%
Physical Form Flakes
Chemical Name or Material 1, 2, 4-Benzenetricarboxylic anhydride, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.