missing translation for 'onlineSavingsMsg'
Learn More

1,2,4-Trimethyl 13C6-benzene (1 mg/mL in Acetonitrile), TRC

Product Code. 30457270
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30457270 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30457270 Supplier Toronto Research Chemicals Supplier No. TRCT7961741ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,2,4-Trimethyl 13C6-benzene
Solvent or Matrix Acetonitrile
Molecular Formula C313C6H12
Quantity 1 mL
Synonym Pseudocumene-13C6
InChI Formula InChI=1S/C9H12/c1-7-4-5-8(2)9(3)6-7/h4-6H,1-3H3/i4+1,5+1,6+1,7+1,8+1,9+1
SMILES C[13C]1=[13CH][13CH]=[13C](C)[13CH]=[13C]1C
IUPAC Name 1,2,4-trimethylbenzene-1,2,3,4,5,6-13C6
Molecular Weight (g/mol) 126.15
Formula Weight 126.11
Concentration or Composition (by Analyte or Components) 1 mg/mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.