missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis[p-(2-hydroxyethoxycarbonyl)benzoyloxy]ethane, TRC

Product Code. 30137830
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137830 10 mg 10mg
30137542 25 mg 25mg
30012904 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137830 Supplier Toronto Research Chemicals Supplier No. TRCB61955010MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2144-69-6
Molecular Formula C22 H22 O10
Molecular Weight (g/mol) 446.404
Synonym 1,4-Benzenedicarboxylic acid, 1,2-ethanediyl bis(2-hydroxyethyl) ester (9CI), Ethylene glycol, terephthalate (3:2) (8CI), Terephthalic acid, ester with ethylene glycol (2:3) (8CI), Terephthalic acid, ethylene bis(2-hydroxyethyl) ester (6CI, 7CI), 1,2-Bis[
IUPAC Name 1-O-[2-[4-(2-hydroxyethoxycarbonyl)benzoyl]oxyethyl] 4-O-(2-hydroxyethyl) benzene-1,4-dicarboxylate
SMILES OCCOC(=O)c1ccc(cc1)C(=O)OCCOC(=O)c2ccc(cc2)C(=O)OCCO

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C22H22O10/c23-9-11-29-19(25)15-1-5-17(6-2-15)21(27)31-13-14-32-22(28)18-7-3-16(4-8-18)20(26)30-12-10-24/h1-8,23-24H,9-14H2
Formula Weight 446.121
Percent Purity >95%
Chemical Name or Material Ethylene Bis(2-hydroxyethyl Terephthalate)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.