missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dehydro Reticuline-d3 Iodide, TRC

Product Code. 30361528
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30361528 2.5 mg 2.5mg
30353785 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30361528 Supplier Toronto Research Chemicals Supplier No. TRCD2300672.5MG
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Spezifikation

Chemical Name or Material 1,2-Dehydro Reticuline-d3 Iodide
Molecular Formula C19H19D3INO4
Quantity 2.5 mg
Synonym 3,4-Dihydro-7-hydroxy-1-(3-hydroxy-4-methoxybenzyl)-6-methoxy-2-methylisoquinolinium-d3 Iodide, 1,2-Dehydroreticuline-d3 Iodide, 3,4-Dihydro-7-hydroxy-1-[(3-hydroxy-4-methoxyphenyl)methyl]-6-methoxy-2-methylisoquinolinium-d3 Iodide
InChI Formula InChI=1S/C19H21NO4.HI/c1-20-7-6-13-10-19(24-3)17(22)11-14(13)15(20)8-12-4-5-18(23-2)16(21)9-12;/h4-5,9-11H,6-8H2,1-3H3,(H-,21,22);1H/i1D3;
SMILES OC1=C(OC)C=CC(CC2=[N+](C([2H])([2H])[2H])CCC3=CC(OC)=C(O)C=C32)=C1.[I-]
Molecular Weight (g/mol) 458.31
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.