Product Code. 30136240
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Unit Size:
100mL
25mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
Quantity
Unit Size
30136240
25 mL
25mL
18072650
100 mL
100mL
18091440
5 mL
5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.
Product Code.
30136240
Supplier
Toronto Research Chemicals
Supplier No.
TRCD22851525ML
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
Chemical Identifiers
CAS
629-14-1
Molecular Formula
C6 H14 O2
Molecular Weight (g/mol)
118.17
Synonym
1,2-Diethoxyethane, Ethane, 1,2-diethoxy-, 1,2-Diethoxyethane, 2-Ethoxyethyl ethyl ether, 3,6-Dioxaoctane, DEE, Diethyl cellosolve, Ethylene diethyl ether, Ethylene glycol diethyl ether, Glycol diethyl ether, Hisolve EME
IUPAC Name
1,2-diethoxyethane
SMILES
CCOCCOCC
Specifications
Quantity
25 mL
InChI Formula
InChI=1S/C6H14O2/c1-3-7-5-6-8-4-2/h3-6H2,1-2H3
Formula Weight
118.0994
Chemical Name or Material
1,2-Diethoxyethane
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our
Privacy Policy .
Spot an opportunity for improvement?