missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Hydroxyethyl) Ester, 1,4-Benzenedicarboxylic Acid, TRC

Product Code. 30008508
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008508 5 mg 5mg
30009606 50 mg 50mg
30011404 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008508 Supplier Toronto Research Chemicals Supplier No. TRCH1137905MG
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1137-99-1
Molecular Formula C10 H10 O5
Molecular Weight (g/mol) 210.183
Synonym 1,4-Benzenedicarboxylic acid, mono(2-hydroxyethyl) ester (9CI), 1-(2-Hydroxyethyl) 1,4-benzenedicarboxylate (ACI), Ethylene glycol, mono(hydrogen terephthalate) (8CI), Terephthalic acid, 2-hydroxyethyl ester (6CI, 7CI), Terephthalic acid, mono(2-hydroxyethyl) ester (8CI), 2-Hydroxyethyl hydrogen terephthalate, 2-Hydroxyethyl terephthalate, Ethylene glycol terephthalate (1:1), Ethylene glycol, hydrogen terephthalate (1:1), Ethylene glycol-terephthalic acid monoester, Hydrogen 2-hydroxyethyl terephthalate, Mono(2-hydroxyethyl) terephthalate, Mono(β-hydroxyethyl) terephthalate, Terephthalic acid ethylene glycol monoester, β-Hydroxyethyl terephthalate, 1,4-Benzenedicarboxylic acid, 1-(2-hydroxyethyl) ester (ACI), Terephthalic acid, mono-2-hydroxyethyl esterMono(2-hydroxyethyl) Ester, 1,4-Benzenedicarboxylic Acid, 2-Hydroxyethyl Ester, Terephthalic Acid, 1-(2-Hydroxyethyl) 1,4-Benzenedicarboxylate, 2-Hydroxyethyl Terephthalate
IUPAC Name 4-(2-hydroxyethoxycarbonyl)benzoic acid
SMILES OCCOC(=O)c1ccc(cc1)C(=O)O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C10H10O5/c11-5-6-15-10(14)8-3-1-7(2-4-8)9(12)13/h1-4,11H,5-6H2,(H,12,13)
Formula Weight 210.053
Chemical Name or Material Terephthalic Acid, Mono-2-hydroxyethyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.