missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Thenoyl)-3,3,3-trifluoroacetone, 99% (dry wt.) may cont. up to ca 2% water

Product Code. 11449742
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11449742 100 g 100g
11439742 25 g 25g
11459742 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11449742 Supplier Thermo Scientific Alfa Aesar Supplier No. A10808.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 326-91-0 | C8H5F3O2S | 222.18 g/mol

1-(2-Thenoyl)-3,3,3-trifluoroacetone is widely used in the analysis of metals such as actinides and lanthanides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(2-Thenoyl)-3,3,3-trifluoroacetone is widely used in the analysis of metals such as actinides and lanthanides.

Solubility
Soluble in methanol and ethanol. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 326-91-0
Molecular Formula C8H5F3O2S
Molecular Weight (g/mol) 222.18
MDL Number MFCD00005445
InChI Key TXBBUSUXYMIVOS-UHFFFAOYSA-N
Synonym 2-thenoyltrifluoroacetone, thenoyltrifluoroacetone, 1,3-butanedione, 4,4,4-trifluoro-1-2-thienyl, 4,4,4-trifluoro-1-thiophen-2-yl butane-1,3-dione, 4,4,4-trifluoro-1-2-thienyl-1,3-butanedione, 4,4,4-trifluoro-1-2-thienyl butane-1,3-dione, perfluoroacetyl 2-thenoyl methane, alpha-thenoyltrifluoroacetone, 1-thenoyl-3,3,3-trifluoroacetone, .alpha.-thenoyltrifluoroacetone
PubChem CID 5601
IUPAC Name 4,4,4-trifluoro-1-thiophen-2-ylbutane-1,3-dione
SMILES FC(F)(F)C(=O)CC(=O)C1=CC=CS1

Specifications

Melting Point 41°C to 44°C
Boiling Point 96°C to 98°C (8 mmHg)
Flash Point 111°C (231°F)
Quantity 100 g
Beilstein 168645
Solubility Information Soluble in methanol and ethanol. Insoluble in water.
Formula Weight 222.18
Percent Purity 99%
Concentration or Composition (by Analyte or Components) May contain up to ≈ 2% water
Assay (dry wt.)
Chemical Name or Material 1- (2-Thenoyl)-3,3,3-trifluoroacetone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.