missing translation for 'onlineSavingsMsg'
Learn More

1,4-Benzenedithiol, 97%

Product Code. 11378736
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11378736 5 g 5g
11388736 250 mg 250mg
11368736 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11378736 Supplier Thermo Scientific Alfa Aesar Supplier No. L19440.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 624-39-5 | C6H6S2 | 142.23 g/mol

1,4-Benzenedithiol is used in single molecule junctions in electronic devices. It is also used to treat silver nano particles and in the preparation of gold electrodes. It finds an application in nanomaterial synthesis and in self-assembled monolayer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Benzenedithiol is used in single molecule junctions in electronic devices. It is also used to treat silver nano particles and in the preparation of gold electrodes. It finds an application in nanomaterial synthesis and in self-assembled monolayer.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 624-39-5
Molecular Formula C6H6S2
Molecular Weight (g/mol) 142.23
MDL Number MFCD00142541
InChI Key WYLQRHZSKIDFEP-UHFFFAOYSA-N
Synonym 1,4-benzenedithiol, dithiohydroquinone, 1,4-dimercaptobenzene, 4-mercaptothiophenol, 1,4-benzene dithiol, pubchem6866, acmc-1b45q, benzene-1,4-dithiol gc
PubChem CID 4691729
IUPAC Name benzene-1,4-dithiol
SMILES SC1=CC=C(S)C=C1

Specifications

Melting Point 95°C to 99°C
Odor Stench
Quantity 5 g
UN Number UN3335
Beilstein 1680022
Sensitivity Air sensitive
Solubility Information Soluble in water.
Formula Weight 142.24
Percent Purity 97%
Chemical Name or Material 1,4-Benzenedithiol
compliance-icons compliance-icons
Product Identifier
  • 1,4-Benzenedithiol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 1,4-Benzenedithiol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.