missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Dichlorobutane-d8, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 1,4-Dichlorobutane-d8 |
| CAS | 83547-96-0 |
| Molecular Formula | C4 D8 Cl2 |
| Quantity | 0.5 g |
| Synonym | Butane-1,1,2,2,3,3,4,4-d8, 1,4-dichloro- (9 CI), 1,4-Dichlorobutane-d8, 1,4-Dichlorobutane-1,1,2,2,3,3,4,4-d8, 1,4-Dichlorobutane-D8, 4-Chlorobutyl-d8 chloride |
| InChI Formula | InChI=1 S/C4H8Cl2/c5-3-1-2-4-6/h1-4H2/i1D2,2D2,3D2,4D2 |
| SMILES | [2 H]C([2 H])(Cl)C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])Cl |
| IUPAC Name | 1,4-dichloro-1,1,2,2,3,3,4,4-octadeuteriobutane |
| Molecular Weight (g/mol) | 135.062 |
| Formula Weight | 134.051 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction