missing translation for 'onlineSavingsMsg'
Learn More

1-Benzhydrylpiperazine, TRC

Product Code. 30138144
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138144 5 g 5g
18061240 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30138144 Supplier Toronto Research Chemicals Supplier No. TRCB1969505G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 841-77-0
Molecular Formula C17 H20 N2
Molecular Weight (g/mol) 252.35
Synonym Piperazine, 1-(diphenylmethyl)-, 1-(Diphenylmethyl)piperazine, 1-Benzhydrylpiperazine, 4-(Diphenylmethyl)piperazine, 4-Benzhydrylpiperazine, Benzhydrylpiperazine, N-(Diphenylmethyl)piperazine, N-Benzhydrylpiperazine, NSC 35536, Norcyclizine
IUPAC Name 1-benzhydrylpiperazine
SMILES C1CN(CCN1)C(c2ccccc2)c3ccccc3

Specifications

Quantity 5 g
InChI Formula InChI=1S/C17H20N2/c1-3-7-15(8-4-1)17(16-9-5-2-6-10-16)19-13-11-18-12-14-19/h1-10,17-18H,11-14H2
Formula Weight 252.1626
Chemical Name or Material 1-(Diphenylmethyl)piperazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.