missing translation for 'onlineSavingsMsg'
Learn More

1-Bis(4-fluorophenyl)methyl Piperazine 97%, Thermo Scientific™

Product Code. 10366060 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10366060 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10366060 Supplier Thermo Scientific Supplier No. 100180100

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

 

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 27469-60-9
Melting Point 89°C to 94°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C17H18F2N2
MDL Number MFCD00038660
Quantity 10 g
Synonym 1-bis 4-fluorophenyl methyl piperazine, 1-bis-4-fluorophenyl methyl piperazine, 1-4,4'-difluorobenzhydryl piperazine, 4,4'-difluorobenzhydrylpiperazine, 1-bis 4-fluorophenyl methylpiperazine, 1 bis 4-fluorophenyl methyl-piperazine, piperazine, 1-bis 4-fluorophenyl methyl, 4,4'-difluorobenzhydryl piperazine
Solubility Information Solubility in water: 0.34g/L (20°C). Other solubilities: >500g/L in methanol, ethanol, 2-propanol, dmf,, acetone and dichloromethane
InChI Key TTXIFFYPVGWLSE-UHFFFAOYSA-N
SMILES C1CN(CCN1)C(C2=CC=C(C=C2)F)C3=CC=C(C=C3)F
IUPAC Name 1-[bis(4-fluorophenyl)methyl]piperazine
Molecular Weight (g/mol) 288.342
PubChem CID 152932
Formula Weight 288.34
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 1-Bis(4-fluorophenyl)methyl piperazine, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 1-Bis(4-fluorophenyl)methyl piperazine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.