missing translation for 'onlineSavingsMsg'
Learn More

1-Bis(4-fluorophenyl)methyl Piperazine, TRC

Product Code. 30134017
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134017 10 g 10g
30133641 25 g 25g
30040767 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134017 Supplier Toronto Research Chemicals Supplier No. TRCB43487510G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27469-60-9
Molecular Formula C17 H18 F2 N2
Molecular Weight (g/mol) 288.34
Synonym 1-[Bis(4-fluorophenyl)methyl]piperazine, Flunarizine Dihydrochloride Imp. A (EP), Piperazine, 1-[bis(4-fluorophenyl)methyl]-, Piperazine, 1-[bis(p-fluorophenyl)methyl]- (8CI), 1-(Di-p-fluorobenzhydryl)piperazine, 4-[Bis(4-fluorophenyl)methyl]piperazine, N-(4,4'-Difluorobenzhydryl)piperazine, N-(Bis(4-fluorophenyl)methyl)piperazine, R 19352, T 841
IUPAC Name 1-[bis(4-fluorophenyl)methyl]piperazine
SMILES Fc1ccc(cc1)C(N2CCNCC2)c3ccc(F)cc3

Specifications

Quantity 10 g
InChI Formula InChI=1S/C17H18F2N2/c18-15-5-1-13(2-6-15)17(21-11-9-20-10-12-21)14-3-7-16(19)8-4-14/h1-8,17,20H,9-12H2
Formula Weight 288.1438
Chemical Name or Material 1-[Bis(4-fluorophenyl)methyl]piperazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.