missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-D-nipecotic acid, 97%

Product Code. 15409147
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15409147 25 g 25g
15499137 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15409147 Supplier Thermo Scientific Alfa Aesar Supplier No. H52416.14
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 163438-09-3 | C11H18NO4 | 228.27 g/mol

1-Boc-D-nipecotic acid is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Boc-D-nipecotic acid is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 163438-09-3
Molecular Formula C11H18NO4
Molecular Weight (g/mol) 228.27
MDL Number MFCD02179173
InChI Key NXILIHONWRXHFA-MRVPVSSYSA-M
Synonym r-boc-nipecotic acid, r-1-tert-butoxycarbonyl piperidine-3-carboxylic acid, r-n-boc-piperidine-3-carboxylic acid, boc-d-nip-oh, r-1-boc-piperidine-3-carboxylic acid, r-1-boc-nipecotic acid, d-1-boc-nipecotic acid, n-boc-r-nipecotic acid, r-boc-nip, r-boc-nip-oh
PubChem CID 643516
SMILES CC(C)(C)OC(=O)N1CCC[C@H](C1)C([O-])=O

Specifications

Quantity 25 g
Beilstein 5539297
IUPAC Name (3R)-1-[(2-methylpropan-2-yl)oxycarbonyl]piperidine-3-carboxylic acid
Formula Weight 229.28
Percent Purity 97%
Chemical Name or Material 1-Boc-D-nipecotic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.