missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-n-butylbenzene, 98%

Product Code. 11436866
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11436866 25 g 25g
11446866 100 g 100g
11456866 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11436866 Supplier Thermo Scientific Alfa Aesar Supplier No. B23979.14
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 41492-05-1 | C10H13Br | 213.118 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41492-05-1
Molecular Formula C10H13Br
Molecular Weight (g/mol) 213.118
MDL Number MFCD00040934
InChI Key BRGVKVZXDWGJBX-UHFFFAOYSA-N
Synonym 1-bromo-4-n-butylbenzene, 4-bromo-n-butylbenzene, 4-n-butylbromobenzene, 1-4-bromophenyl butane, benzene, 1-bromo-4-butyl, 4-butyl bromobenzene, 4-butyl-bromobenzene, 4-1-butylbromobenzene, 4-butyl phenylbromide, pubchem15109
PubChem CID 521059
IUPAC Name 1-bromo-4-butylbenzene
SMILES CCCCC1=CC=C(C=C1)Br

Specifications

Density 1.208
Boiling Point 242°C
Flash Point 108°C (226°F)
Refractive Index 1.53
Quantity 25 g
Beilstein 2080263
Formula Weight 213.12
Percent Purity 98%
Chemical Name or Material 1-Bromo-4-n-butylbenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.