missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromopentane-4,4,5,5,5-d5, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | 1-Bromopentane-4,4,5,5,5-D5 |
| CAS | 83418-34-2 |
| Molecular Formula | C5 D5 H6 Br |
| Quantity | 0.5 g |
| Synonym | Pentane-1,1,1,2,2-d5, 5-bromo- (9 CI), 1-Bromopentane-4,4,5,5,5-d5, 5-Bromopentane-1,1,1,2,2-d5, 1-Bromo-4,4,5,5,5-pentadeuteriopentane, 1-Bromopentane-d5, 1-Bromopentane-4,4,5,5,5-D5 |
| InChI Formula | InChI=1 S/C5H11Br/c1-2-3-4-5-6/h2-5H2,1H3/i1D3,2D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])CCCBr |
| IUPAC Name | 5-bromo-1,1,1,2,2-pentadeuteriopentane |
| Molecular Weight (g/mol) | 156.0757 |
| Formula Weight | 155.0358 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction