missing translation for 'onlineSavingsMsg'
Learn More

1-Bromotetradecane, TRC

Product Code. 30240607
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30240607 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30240607 Supplier Toronto Research Chemicals Supplier No. TRCB6880152.5G
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-71-0
Molecular Formula C14H29Br
Molecular Weight (g/mol) 277.28
Synonym 1-Bromotetradecane, 1-Tetradecyl bromide, Myristyl bromide, NSC 83468, Tetradecyl bromide, n-Tetradecyl bromide, n-Tetradecyl-1-bromide, 1-Tetradecyl Bromide, Myristyl Bromide, NSC 83468, Tetradecyl Bromide, n-Tetradecyl Bromide, n-Tetradecyl-1-bromide
IUPAC Name 1-bromotetradecane
SMILES CCCCCCCCCCCCCCBr

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C14H29Br/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15/h2-14H2,1H3
Formula Weight 276.1453
Chemical Name or Material 1-Bromotetradecane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.