missing translation for 'onlineSavingsMsg'
Learn More

1-Butyl-2,3-dimethylimidazolium chloride, 99%

Product Code. p-7029770
Change view
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11332309 5 g 5g
11342309 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11332309 Supplier Thermo Scientific Alfa Aesar Supplier No. H27270.06
Add to basket
This item is not returnable. View return policy

CAS: 98892-75-2 | C9H17ClN2 | 188.699 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98892-75-2
Molecular Formula C9H17ClN2
Molecular Weight (g/mol) 188.699
MDL Number MFCD03093291
InChI Key HHHYPTORQNESCU-UHFFFAOYSA-M
Synonym 1-butyl-2,3-dimethylimidazolium chloride, 1-butyl-2,3-dimethyl-1h-imidazol-3-ium chloride, 1-butyl-2,3-dimethylimidazol-1-ium chloride, 3-butyl-1,2-dimethylimidazol-1-ium chloride, acmc-209sba, dsstox_cid_27882, dsstox_rid_82635, dsstox_gsid_47906, 1-butyl-2,3-dimethylimidazoliumchloride, 1-butyl-2,3-dime-thylimidazolium chloride
PubChem CID 2734167
IUPAC Name 1-butyl-2,3-dimethylimidazol-3-ium;chloride
SMILES CCCCN1C=C[N+](=C1C)C.[Cl-]

Specifications

Melting Point 89.5°C to 93.2°C
Quantity 5 g
Formula Weight 188.69
Percent Purity 99%
Chemical Name or Material 1-Butyl-2,3-dimethylimidazolium chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.