missing translation for 'onlineSavingsMsg'
Learn More

1-Chloropinacolone, 95%, Thermo Scientific™

Product Code. 11487897
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11487897 25 g 25g
11497897 100 g 100g
11407907 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11487897 Supplier Thermo Scientific Alfa Aesar Supplier No. A16576.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
1-Chloropinacolone is used as an intermediate for triazole compounds ( a five-membered ring structure compound contains three nitrogens and two carbons in the ring) which have biological activities such as antiviral, antibacterial, antifungal and antituberculous.

Solubility
Not miscible in water.

Notes
Incompatible with oxidizing agents and bases. Store in a cool, dry, conditions in well sealed container. Store at 4°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13547-70-1
Molecular Formula C6H11ClO
Molecular Weight (g/mol) 134.603
MDL Number MFCD00035688
InChI Key ULSAJQMHTGKPIY-UHFFFAOYSA-N
Synonym 1-chloropinacolone, 1-chloro-3,3-dimethyl-2-butanone, chlorpinakolin, 2-butanone, 1-chloro-3,3-dimethyl, 1-monochloropinacoline, tert-butyl chloromethyl ketone, alpha-chloropinacolin, alpha-chloropinacoline, chloromethyl tert-butyl ketone, chloropinacoline
PubChem CID 83572
IUPAC Name 1-chloro-3,3-dimethylbutan-2-one
SMILES CC(C)(C)C(=O)CCl

Specifications

Melting Point -13°C
Boiling Point 170°C to 173°C
Flash Point 67°C (153°F)
Odor Pungent
Linear Formula (CH3)3CCOCH2Cl
Quantity 25 g
UN Number UN3382
Formula Weight 134.59
Percent Purity 95%
Chemical Name or Material 1-Chloropinacolone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.