missing translation for 'onlineSavingsMsg'
Learn More

1-Cyclopentenecarbonitrile, 95%

Product Code. 11465560
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11465560 1 g 1g
11475560 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11465560 Supplier Thermo Scientific Alfa Aesar Supplier No. H25836.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3047-38-9 | C6H7N | 93.129 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3047-38-9
Molecular Formula C6H7N
Molecular Weight (g/mol) 93.129
MDL Number MFCD00043765
InChI Key XBVZRFXCDCYXAX-UHFFFAOYSA-N
Synonym 1-cyanocyclopentene, cyclopent-1-enecarbonitrile, 1-cyclopentenecarbonitrile, 1-cyclopentene-1-carbonitrile, cyclopent-1-ene-1-carbonitrile, 1-cyanocyclopenten, acmc-20aovl, 1-cyclopentenyl cyanide, 1-cyano-1-cyclopentene
PubChem CID 137808
IUPAC Name cyclopentene-1-carbonitrile
SMILES C1CC=C(C1)C#N

Specifications

Density 0.944
Boiling Point 72°C (15 mmHg)
Refractive Index 1.4715
Quantity 1 g
UN Number UN3276
Formula Weight 93.14
Percent Purity 97%
Chemical Name or Material 1-Cyclopentenecarbonitrile
compliance-icons
Product Identifier
  • 1-Cyclopentenecarbonitrile
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 1-Cyclopentenecarbonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.