missing translation for 'onlineSavingsMsg'
Learn More

(1-Ethoxycarbonylethylidene)triphenylphosphorane, 97%

Product Code. 11417507
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
250 g
Unit Size:
10g
250g
25g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11417507 10 g 10g
11464180 25 g 25g
11427507 50 g 50g
11437507 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11417507 Supplier Thermo Scientific Alfa Aesar Supplier No. A15619.09
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 5717-37-3 | C23H23O2P | 362.41 g/mol

(1-Ethoxycarbonylethylidene)triphenylphosphorane is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals. It’s also a Horner-Wadsworth-Emmons reagent. It can be used for olefination. It can also be utilized for the synthesis of more complex compounds, such as (29S,37S)-isomer of malevamide E, a potent ion-channel inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1-Ethoxycarbonylethylidene)triphenylphosphorane is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals. It’s also a Horner-Wadsworth-Emmons reagent. It can be used for olefination. It can also be utilized for the synthesis of more complex compounds, such as (29S,37S)-isomer of malevamide E, a potent ion-channel inhibitor.

Solubility
Soluble in water.

Notes
Air sensitive. Avoid contact with oxygen and air. Store away from oxidizing agent. Contains triphenylphosphine oxide.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5717-37-3
Molecular Formula C23H23O2P
Molecular Weight (g/mol) 362.41
MDL Number MFCD00009160
InChI Key KZENFXVDPUMQOE-UHFFFAOYSA-N
Synonym ethyl 2-triphenylphosphoranylidene propanoate, carbethoxyethylidene triphenylphosphorane, ethyl 2-triphenylphosphoranylidene propionate, 1-ethoxycarbonylethylidene triphenylphosphorane, propanoic acid, 2-triphenylphosphoranylidene-, ethyl ester, 1-ethoxycarbonyl ethylidenetriphenylphosphorane, ethyl 2-triphenyl-lambda5-phosphanylidene propanoate, ethyl 2-triphenyl-$l^ 5-phosphanylidene propanoate, 1-ethoxycarbonylethylethylidene triphenylphosporane, carbethoxyethylidine triphenylphosphorane
PubChem CID 79792
IUPAC Name ethyl 2-(triphenyl-$l^{5}-phosphanylidene)propanoate
SMILES CCOC(=O)C(C)=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 154°C to 160°C
Quantity 10 g
Beilstein 757112
Sensitivity Air sensitive
Solubility Information Soluble in water.
Formula Weight 362.41
Percent Purity 97%
Chemical Name or Material (1-Ethoxycarbonylethylidene)triphenylphosphorane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.