Learn More
1-Ethyl-3-methylimidazolium trifluoromethanesulfonate, 98+%
Product Code. 11309715
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11309715 2 g 2g
11319715 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11309715 Supplier Thermo Scientific Alfa Aesar Supplier No. L19764.04
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 145022-44-2 | C7H11F3N2O3S | 260.231 g/mol
Recently used in the preparation of hydrophobic, highly conductive ambient-temperature molten salts, and in the preparation of dual intercalating molten electrolyte batteries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Recently used in the preparation of hydrophobic, highly conductive ambient-temperature molten salts, and in the preparation of dual intercalating molten electrolyte batteries.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 145022-44-2
Molecular Formula C7H11F3N2O3S
Molecular Weight (g/mol) 260.231
MDL Number MFCD00210008
InChI Key ZPTRYWVRCNOTAS-UHFFFAOYSA-M
Synonym 1-ethyl-3-methylimidazolium trifluoromethanesulfonate, 1-ethyl-3-methylimidazolium triflate, 1-ethyl-3-methyl-1h-imidazol-3-ium trifluoromethanesulfonate, acmc-209ctv, dsstox_cid_27941, dsstox_rid_82693, dsstox_gsid_47965, 1-ethyl-3-methylimidazoliumtrifluoromethanesulfonate, ksc685i3d, 1-ethyl-3-methylimidazolium trifluoromethanesulfon
PubChem CID 2758874
IUPAC Name 1-ethyl-3-methylimidazol-3-ium;trifluoromethanesulfonate
SMILES CCN1C=C[N+](=C1)C.C(F)(F)(F)S(=O)(=O)[O-]
Specifications
Density 1.387
Flash Point None
Refractive Index 1.435
Quantity 2 g
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 260.24
Percent Purity ≥98%
Chemical Name or Material 1-Ethyl-3-methylimidazolium trifluoromethanesulfonate