missing translation for 'onlineSavingsMsg'
Learn More

1-Methoxy-2-propanol, 99+%

Product Code. 11394298
Change view
Click to view available options
Quantity:
250 mL
1 L
4 L
4 x 4 L
Unit Size:
1 L
16 L
250mL
4 L
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11394298 250 mL 250mL
11353316 250 mL 250mL
11304308 1 L 1 L
11363316 1 L 1 L
11314308 4 L 4 L
11324308 4 x 4 L 16 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 11394298 Supplier Thermo Scientific Alfa Aesar Supplier No. 041547.AK
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 107-98-2 | C4H10O2 | 90.12 g/mol

1-Methoxy-2-propanol is used as a reagent in the synthesis of 2-amino-3-carboxy-4-phenylthiophenes, which acts as a protein kinase C inhibitors. It is also used as a reagent in the synthesis of metolachlor. It acts as a good biological indicator. Further, it is used as a solvent and an antifreeze agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-98-2
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.12
MDL Number MFCD00004537
InChI Key ARXJGSRGQADJSQ-UHFFFAOYNA-N
Synonym 1-methoxy-2-propanol, pgme, methoxyisopropanol, 2-propanol, 1-methoxy, closol, dowtherm 209, propylene glycol monomethyl ether, poly-solve mpm, propasol solvent m, dowanol 33b
PubChem CID 7900
IUPAC Name 1-methoxypropan-2-ol
SMILES COCC(C)O

Specifications

Melting Point -100°C
Color Colorless
Density 0.926 g/mL
pH 4 to 7
Boiling Point 119°C to 120°C
Flash Point 35°C (95°F)
Assay Percent Range >99%
Odor Alcohol-like
Refractive Index 1.403
Quantity 250 mL
UN Number UN3092
Beilstein 1731270
Solubility Information Miscible with water,methanol and ether.
Formula Weight 90.12
Percent Purity ≥99%
Physical Form Liquid
Chemical Name or Material 1-Methoxy-2-propanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.