missing translation for 'onlineSavingsMsg'
Learn More

1-Methylpyrrole, 99%

Product Code. 11439447
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Unit Size:
100mL
2500mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11439447 100 mL 100mL
11449447 500 mL 500mL
11456964 2500 mL 2500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11439447 Supplier Thermo Scientific Alfa Aesar Supplier No. B21451.AE
  • ONLINE EXCLUSIVE PRICE
    72.80€
    65.65€ / 100mL
    Save 7.15€
    9% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 96-54-8 | C5H7N | 81.118 g/mol

1-Methylpyrrole, can be used as organic synthetic materials, widely used in medicine. It is also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Methylpyrrole, can be used as organic synthetic materials, widely used in medicine. It is also used as a pharmaceutical intermediate.

Solubility
Soluble in organic solvents such as alcohol and so on, but insoluble in water.

Notes
Keep away from acids, acid chlorides, acid anhydrides, strong oxidizing agents, carbon dioxide. Keep away from acids, acid chlorides, acid anhydrides, strong oxidizing agents, carbon dioxide. Stable under normal temperatures and pressures.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-54-8
Molecular Formula C5H7N
Molecular Weight (g/mol) 81.118
MDL Number MFCD00005345
InChI Key OXHNLMTVIGZXSG-UHFFFAOYSA-N
Synonym 1-methyl-1h-pyrrole, n-methylpyrrole, n-methyl pyrrole, 1h-pyrrole, 1-methyl, pyrrole, 1-methyl, methylpyrrole, n-methylpyrrol, 1-methylpyrrol, unii-ng5gpn98zg, ccris 2934
PubChem CID 7304
IUPAC Name 1-methylpyrrole
SMILES CN1C=CC=C1

Specifications

Melting Point -57°C
Density 0.912
Boiling Point 112°C to 113°C
Flash Point 15°C (59°F)
Refractive Index 1.488
Quantity 100 mL
UN Number UN1993
Beilstein 104181
Solubility Information Soluble in organic solvents such as alcohol and so on,but insoluble in water.
Formula Weight 81.12
Percent Purity 99%
Chemical Name or Material 1-Methylpyrrole
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.