missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-1-pentyn-3-ol, 97%

Product Code. 15410208
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15410208 1 g 1g
15420208 5 g 5g
15430208 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15410208 Supplier Thermo Scientific Alfa Aesar Supplier No. H53416.03
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 27975-78-6 | C11H12O | 160.216 g/mol

Reactant in the heterogeneously catalyzed etherification of glycerol, also in fritsch-wiechell rearrangement of 1-chlorovinyl sulfoxides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant in the heterogeneously catalyzed etherification of glycerol, also in fritsch-wiechell rearrangement of 1-chlorovinyl sulfoxides.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27975-78-6
Molecular Formula C11H12O
Molecular Weight (g/mol) 160.216
MDL Number MFCD00041589
InChI Key QWCMSASONHVIHV-UHFFFAOYSA-N
Synonym 1-phenyl-1-pentyn-3-ol, 1-pentyn-3-ol,1-phenyl, 1-pentyn-3-ol, 1-phenyl-, s, acmc-20mqra, 1-pentyn-3-ol, 1-phenyl
PubChem CID 11789508
IUPAC Name 1-phenylpent-1-yn-3-ol
SMILES CCC(C#CC1=CC=CC=C1)O

Specifications

Density 1.336
Flash Point >110°C (230°F)
Refractive Index 1.559
Quantity 1 g
Formula Weight 160.22
Percent Purity 97%
Chemical Name or Material 1-Phenyl-1-pentyn-3-ol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.