missing translation for 'onlineSavingsMsg'
Learn More

1-Propanol, TRC

Product Code. 30137960
Change view
Click to view available options
Quantity:
100 mL
250 mL
500 mL
Unit Size:
100mL
250mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137960 500 mL 500mL
30137969 250 mL 250mL
17992875 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.
Product Code. 30137960 Supplier Toronto Research Chemicals Supplier No. TRCP760860500ML

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71-23-8
Molecular Formula C3 H8 O
Molecular Weight (g/mol) 60.1
Synonym 1-Propanol (9CI, ACI), Propyl alcohol (8CI), 1-Hydroxypropane, Ethylcarbinol, n-Propanol, n-Propyl alcohol, NSC 30300, Optal, Osmosol extra, Propanol, Propan-1-ol
IUPAC Name propan-1-ol
SMILES CCCO

Specifications

Quantity 500 mL
InChI Formula InChI=1S/C3H8O/c1-2-3-4/h4H,2-3H2,1H3
Formula Weight 60.0575
Chemical Name or Material 1-Propanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.