Learn More
10,12-Docosadiynedioic acid, 95%
Product Code. p-7045423
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11342926 1 g 1g
11352926 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11342926 Supplier Thermo Scientific Alfa Aesar Supplier No. L10647.03
Add to basket
missing translation for 'validMainframeMsg'
CAS: 28393-02-4 | C22H34O4 | 362.51 g/mol
Polymerizable diacid (10,12-docosadiynedioic acid) was used as obtained from GFS Chemicals. Synthesis of dimesogenic liquid crystals with both cholesterol and azobenzene groups is by the esterification of 10,12-docosadiynedioic acid. .
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Polymerizable diacid (10,12-docosadiynedioic acid) was used as obtained from GFS Chemicals. Synthesis of dimesogenic liquid crystals with both cholesterol and azobenzene groups is by the esterification of 10,12-docosadiynedioic acid. .
Solubility
Insoluble in water.
Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 28393-02-4
Molecular Formula C22H34O4
Molecular Weight (g/mol) 362.51
MDL Number MFCD00078306
InChI Key XCUAGNLOPZWTEH-UHFFFAOYSA-N
Synonym 10,12-docosadiynedioic acid, 10,12-docosadiyndioic acid, 10,12-docosadiynedioicacid, acmc-20aozt, 10,12-docasadiyndioic acid, 10,12-docosadiynedioic acid #, icosa-9,11-diyne-1,20-dicarboxylic acid
PubChem CID 544138
IUPAC Name docosa-10,12-diynedioic acid
SMILES C(CCCCC(=O)O)CCCC#CC#CCCCCCCCCC(=O)O
Specifications
Melting Point 110°C to 112°C
Quantity 1 g
Beilstein 1803395
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 362.51
Percent Purity 95%
Chemical Name or Material 10,12-Docosadiynedioic acid