missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Propyl Ether, TRC

Product Code. 17973035
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17973035 100 mg 100mg
30135498 500 mg 500mg
30137396 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17973035 Supplier Toronto Research Chemicals Supplier No. TRCI748500100MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 627-08-7
Molecular Formula C6 H14 O
Molecular Weight (g/mol) 102.175
Synonym 1-(1-Methylethoxy)propane, 1-(Propan-2-yloxy)propane, Isopropyl propyl ether, Propyl iso-propyl ether, Propyl isopropyl ether;
IUPAC Name 1-propan-2-yloxypropane
SMILES CCCOC(C)C

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C6H14O/c1-4-5-7-6(2)3/h6H,4-5H2,1-3H3
Formula Weight 102.104
Chemical Name or Material Isopropyl Propyl Ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.