missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Exosome Component 9 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB11616-100
This item is not returnable.
View return policy
Description
Exosome Component 9 Monoclonal antibody specifically detects Exosome Component 9 in Human samples. It is validated for Immunohistochemistry,Immunocytochemistry
Specifications
| Exosome Component 9 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. | |
| Autoantigen PM/Scl 1, EC 3.1.13, exosome complex component RRP45, exosome complex exonuclease RRP45, P5, p6, P75 polymyositis-scleroderma overlap syndrome associated autoantigen, PCH1D, PM/Scl-75, PMSCL autoantigen, 75kD, PMSCL1, Polymyositis/scleroderma autoantigen 1, polymyositis/scleroderma autoantigen 1 (75kD), polymyositis/scleroderma autoantigen 1, 75kDa, RRP45, Rrp45p | |
| Synthetic Peptide, Accession # Q06265 | |
| 100 μg | |
| Primary | |
| Reconstitute lyophilized material at 0.2 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20°C to -70 °C under sterile conditions after reconstitution. | |
| IgG1 |
| Immunohistochemistry, Immunocytochemistry | |
| 1090209 | |
| Immunohistochemistry 3-25 μg/mL, Immunocytochemistry 3-25 μg/mL | |
| Q06265 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5393 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction