missing translation for 'onlineSavingsMsg'
Learn More
Learn More
14-3-3 sigma Mouse anti-Human, Clone: 3F10D9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66251-1-IG-20UL
This item is not returnable.
View return policy
Description
This gene encodes a cell cycle checkpoint protein. The encoded protein binds to translation and initiation factors and functions as a regulator of mitotic translation. In response to DNA damage this protein plays a role in preventing DNA errors during mitosis.Specifications
| 14-3-3 sigma | |
| Monoclonal | |
| 1.37 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P31947 | |
| SFN | |
| 14-3-3 sigma Fusion Protein Ag12644 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot, Immunocytochemistry | |
| 3F10D9 | |
| Unconjugated | |
| SFN | |
| 14 3 3 protein sigma, HME1, SFN, stratifin, YWHAS | |
| Mouse | |
| Protein G | |
| RUO | |
| 2810 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction