missing translation for 'onlineSavingsMsg'
Learn More

14-3-3 sigma Mouse anti-Human, Clone: 3F10D9, Proteintech

Mouse Monoclonal Antibody

145.00€ - 410.00€

Specifications

Antigen 14-3-3 sigma
Clone 3F10D9
Concentration 1.37 mg/mL
Applications Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot, Immunocytochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16847955
View Documents
Proteintech
66251-1-IG-20UL
20 μL
145.00€
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16837955
View Documents
Proteintech
66251-1-IG-150UL
150 μL
410.00€
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This gene encodes a cell cycle checkpoint protein. The encoded protein binds to translation and initiation factors and functions as a regulator of mitotic translation. In response to DNA damage this protein plays a role in preventing DNA errors during mitosis.
Specifications

Specifications

14-3-3 sigma
1.37 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
14 3 3 protein sigma, HME1, SFN, stratifin, YWHAS
SFN
IgG1
Protein G
Antibody
3F10D9
Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot, Immunocytochemistry
Unconjugated
Mouse
Human
P31947
2810
14-3-3 sigma Fusion Protein Ag12644
Primary
-20°C
SFN
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.