missing translation for 'onlineSavingsMsg'
Learn More

15-Acetyl Deoxynivalenol-13D2 (~90%), TRC

Product Code. 30326541
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30326541 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30326541 Supplier Toronto Research Chemicals Supplier No. TRCA17351810MG
 Restricted Item
Estimated Shipment: 18-03-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 15-Acetyl Deoxynivalenol-13C2
Molecular Formula 13C2 C15 H22 O7
Quantity 10 mg
Synonym (3alpha,7alpha)-15-(Acetyloxy)-12,13-epoxy-3,7-dihydroxy-trichothec-9-en-8-one-13C2, 15-Acetylvomitoxin-13C2, 15-O-Acetyl-4-deoxynivalenol-13C2, Deoxynivalenol 15-Acetate-13C2
InChI Formula InChI=1S/C17H22O7/c1-8-4-11-16(6-22-9(2)18,13(21)12(8)20)15(3)5-10(19)14(24-11)17(15)7-23-17/h4,10-11,13-14,19,21H,5-7H2,1-3H3/t10-,11-,13-,14-,15-,16-,17+/m1/s1/i2+1,9+1
SMILES [13CH3][13C](=O)OC[C@]12[C@H](O)C(=O)C(=C[C@H]1O[C@@H]3[C@H](O)C[C@@]2(C)[C@]34CO4)C
IUPAC Name [(1R,2R,3S,7R,9R,10R,12S)-3,10-dihydroxy-1,5-dimethyl-4-oxospiro[8-oxatricyclo[7.2.1.02,7]dodec-5-ene-12,2'-oxirane]-2-yl]methyl acetate
Molecular Weight (g/mol) 340.338
Formula Weight 340.143
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.