missing translation for 'onlineSavingsMsg'
Learn More
Learn More
17-Beta-Estradiol Antibody (4S24 (BGN/06/8824)) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-62280-0.025MG
This item is not returnable.
View return policy
Description
17-Beta-Estradiol Monoclonal specifically detects 17-Beta-Estradiol in All Species samples. It is validated for ELISA.
Specifications
| 17-Beta-Estradiol | |
| Monoclonal | |
| 1 mg/mL | |
| ELISA 1:100-1:2000 | |
| 17 beta Oestradiol | |
| 17-beta estradiol conjugated to bovine serum albumin at position 6. | |
| 0.025 mg | |
| Primary | |
| All species | |
| IgG1 |
| ELISA | |
| 4S24 (BGN/06/8824) | |
| Unconjugated | |
| PBS with 0.09% Sodium Azide | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| NB100-62280 recognizes 17-beta estradiol, a mammalian estrogenic hormone, produced in the ovaries, placenta and testis that has a role in normal sexual development and the regulation of the menstrual cycle. The following cross reactivites have been observed: 17-beta Estradiol 100% 17-beta Estradiol-BSA 90% Estriol <0.9% Testosterone <0.01% | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction