missing translation for 'onlineSavingsMsg'
Learn More
Learn More
17-Beta-Estradiol Antibody (4S24 (BGN/06/8824)) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
190.00€ - 487.00€
Specifications
| Antigen | 17-Beta-Estradiol |
|---|---|
| Clone | 4S24 (BGN/06/8824) |
| Concentration | 1 mg/mL |
| Dilution | ELISA 1:100-1:2000 |
| Applications | ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18006064
|
Novus Biologicals
NB100-62280-0.025MG |
0.025 mg |
190.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18097175
|
Novus Biologicals
NB100-62280 |
0.1 mg |
487.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
17-Beta-Estradiol Monoclonal specifically detects 17-Beta-Estradiol in All Species samples. It is validated for ELISA.Specifications
| 17-Beta-Estradiol | |
| 1 mg/mL | |
| ELISA | |
| Unconjugated | |
| RUO | |
| 17-beta estradiol conjugated to bovine serum albumin at position 6. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 4S24 (BGN/06/8824) | |
| ELISA 1:100-1:2000 | |
| Monoclonal | |
| Mouse | |
| All species | |
| IgG1 | |
| Affinity Purified | |
| NB100-62280 recognizes 17-beta estradiol, a mammalian estrogenic hormone, produced in the ovaries, placenta and testis that has a role in normal sexual development and the regulation of the menstrual cycle. The following cross reactivites have been observed: 17-beta Estradiol 100% 17-beta Estradiol-BSA 90% Estriol <0.9% Testosterone <0.01% |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title