missing translation for 'onlineSavingsMsg'
Learn More

2-O-(p-Nitrophenyl)-Alpha-D-N-acetylneuraminic Acid, Sodium Salt, X Hydrate, TRC

Product Code. 30136885
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136885 1 mg 1mg
30134584 10 mg 10mg
30009168 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136885 Supplier Toronto Research Chemicals Supplier No. TRCN5025011MG
 Restricted Item
Estimated Shipment: 02-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123549-14-4
Molecular Formula C17 H21 N2 O11 . Na . H2 O
Molecular Weight (g/mol) 470.36
IUPAC Name sodium;(2S,4S,5R,6R)-5-acetamido-4-hydroxy-2-(4-nitrophenoxy)-6-[(1R,2R)-1,2,3-trihydroxypropyl]oxane-2-carboxylate;hydrate
SMILES O.[Na+].CC(=O)N[C@@H]1[C@@H](O)C[C@@](Oc2ccc(cc2)[N+](=O)[O-])(O[C@H]1[C@H](O)[C@H](O)CO)C(=O)[O-]

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C17H22N2O11.Na.H2O/c1-8(21)18-13-11(22)6-17(16(25)26,30-15(13)14(24)12(23)7-20)29-10-4-2-9(3-5-10)19(27)28;;/h2-5,11-15,20,22-24H,6-7H2,1H3,(H,18,21)(H,25,26);;1H2/q;+1;/p-1/t11-,12+,13+,14+,15+,17+;;/m0../s1
Formula Weight 470.1149
Chemical Name or Material 2-O-(p-Nitrophenyl)-alpha-D-N-acetylneuraminic acid sodium salt hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.