missing translation for 'onlineSavingsMsg'
Learn More

2-(1-Naphthyl)ethanol, 95%, Thermo Scientific Chemicals

Product Code. 11322209
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11322209 10 g 10g
11332209 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11322209 Supplier Thermo Scientific Alfa Aesar Supplier No. H27218.09
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 773-99-9
Molecular Formula C12H12O
Molecular Weight (g/mol) 172.227
MDL Number MFCD00004050
InChI Key RXWNCMHRJCOWDK-UHFFFAOYSA-N
Synonym 1-naphthaleneethanol, 2-1-naphthyl ethanol, 2-naphthalen-1-yl ethanol, 1-2-hydroxyethyl naphthalene, naphthalen-1-ethanol, 2-naphthylethanol, 1-naphthyl ethanol, 1-naphthalene ethanol, 2-naphthalen-1-yl ethan-1-ol, 1-napthaleneethanol
PubChem CID 13047
IUPAC Name 2-naphthalen-1-ylethanol
SMILES C1=CC=C2C(=C1)C=CC=C2CCO

Specifications

Melting Point 60°C to 64°C
Boiling Point 186°C (17 mmHg)
Quantity 10 g
Formula Weight 172.23
Percent Purity 95%
Chemical Name or Material 2-(1-Naphthyl)ethanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.