missing translation for 'onlineSavingsMsg'
Learn More

2,2-Bis[4-(2-hydroxy-2-methylethoxy)phenyl]Propane, TRC

Product Code. 30005203
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005203 500 mg 500mg
30003898 100 mg 100mg
30007280 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30005203 Supplier Toronto Research Chemicals Supplier No. TRCB016472500MG
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 116-37-0
Molecular Formula C21H28O4
Molecular Weight (g/mol) 344.44
Synonym 1,1'-[(1-Methylethylidene)bis(4,1-phenyleneoxy)]-bis[2-propanol], 1,1'-Isopropylidenebis[4-(2-hydroxypropoxy)Benzene], 2,2-Bis[4-(2-hydroxypropoxy)phenyl]Propane, Bisphenol A-bis(2-hydroxypropyl) Ether, Dianol 33, Himer BP 2P, Isopropylidenediphenoxypropanol, NSC 408494, Propoxylated Bisphenol A 2 Mole Adduct, Dipropoxylated Bisphenol A
IUPAC Name 1-[4-[2-[4-(2-hydroxypropoxy)phenyl]propan-2-yl]phenoxy]propan-2-ol
SMILES CC(O)COc1ccc(cc1)C(C)(C)c2ccc(OCC(C)O)cc2

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C21H28O4/c1-15(22)13-24-19-9-5-17(6-10-19)21(3,4)18-7-11-20(12-8-18)25-14-16(2)23/h5-12,15-16,22-23H,13-14H2,1-4H3
Formula Weight 344.1988
Percent Purity >95%
Chemical Name or Material 2,2-Bis[4-(2-hydroxy-2-methylethoxy)phenyl]Propane
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.