missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethoxy-2-phenylacetophenone, TRC

Product Code. 17914135
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17914135 10 g 10g
17953625 50 g 50g
17941945 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17914135 Supplier Toronto Research Chemicals Supplier No. TRCD47068010G
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24650-42-8
Molecular Formula C16 H16 O3
Molecular Weight (g/mol) 256.3
Synonym Benzil dimethyl acetal (6CI), 2,2-Dimethoxy-1,2-diphenylethanone, 1,2-Diphenyl-2,2-dimethoxyethanone, 2,2-Dimethoxy-1,2-diphenyl-1-ethanone, 2,2-Dimethoxy-1,2-diphenylethan-1-one, 2,2-Dimethoxy-2-phenylacetophenone, 2,2-Dimethoxyphenylacetophenone, 2-Phenyl-2,2-dimethoxyacetophenone, Aronix C 101, BDK, Benzil dimethyl ketal, Benzil mono(dimethyl acetal), Benzil mono(dimethyl ketal), Benzoin dimethyl ether, C 101, Chemcure BDK, Ciba 651, DMPA, Doublecure BDK, Esacure KB 1, I 651, IC 127, IR 651, IRG 651, Irgacure 621, Irgacure 641, Irgacure 651, Irgacure 654, Irgacure 671, Irgacure 951, Irgacure E 651, Irgacure I 651, KB 1, Kayacure BDMK, Lucirin BDK, Micure BK 6, Omnirad 651, Omnirad BDK, PIBDK, Photocure 51, Photoinitiator 651, Photoinitiator 6512, Photoinitiator BDK, Photomer 51, Quantacure BDK, SB-PI 718, Speedcure BKL, UV 651, α,α-Dimethoxy-α-phenylacetophenone, α,α-Dimethoxydeoxybenzoin, ω,ω-Dimethoxy-ω-phenylacetophenone
IUPAC Name 2,2-dimethoxy-1,2-diphenylethanone
SMILES COC(OC)(C(=O)c1ccccc1)c2ccccc2

Specifications

Quantity 10 g
InChI Formula InChI=1S/C16H16O3/c1-18-16(19-2,14-11-7-4-8-12-14)15(17)13-9-5-3-6-10-13/h3-12H,1-2H3
Formula Weight 256.1099
Chemical Name or Material 2,2-Dimethoxy-2-phenylacetophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.