missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethyl-1,3-dioxan-5-one, TRC

Product Code. 30377103
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30377103 5 g
30369971 1 g
30383981 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30377103 Supplier Toronto Research Chemicals Supplier No. TRCD4728405G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74181-34-3
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.14
Synonym 1,3-Dioxan-5-one, 2,2-dimethyl-, 2,2-Dimethyl-1,3-dioxan-5-one
IUPAC Name 2,2-dimethyl-1,3-dioxan-5-one
SMILES CC1(C)OCC(=O)CO1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C6H10O3/c1-6(2)8-3-5(7)4-9-6/h3-4H2,1-2H3
Formula Weight 130.063
Chemical Name or Material 2,2-Dimethyl-1,3-dioxan-5-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.