missing translation for 'onlineSavingsMsg'
Learn More

2,2-Diphenylpentanoic Acid, TRC

Product Code. 30384630
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30384630 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30384630 Supplier Toronto Research Chemicals Supplier No. TRCD4918351G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 841-32-7
Molecular Formula C17 H18 O2
Quantity 1 g
InChI Formula InChI=1S/C17H18O2/c1-2-13-17(16(18)19,14-9-5-3-6-10-14)15-11-7-4-8-12-15/h3-12H,2,13H2,1H3,(H,18,19)
SMILES CCCC(C(=O)O)(c1ccccc1)c2ccccc2
IUPAC Name 2,2-diphenylpentanoic acid
Molecular Weight (g/mol) 254.32
Formula Weight 254.1307
Chemical Name or Material 2,2-Diphenylpentanoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.