Learn More
2-(2-Methyl-1H-imidazol-1-yl)aniline, 97%, Thermo Scientific™
Product Code. 10787285
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10787285 250 mg Amber Glass Bottle 250mg
10797285 1 g Amber Glass Bottle 1g
10108164 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10787285 Supplier Thermo Scientific Maybridge Supplier No. CC61214CB
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
missing translation for 'validMainframeMsg'
Chemical Identifiers
CAS 26286-55-5
Molecular Formula C10H11N3
Molecular Weight (g/mol) 173.22
MDL Number MFCD08699292
InChI Key LLSZMUSXQITSSJ-UHFFFAOYSA-N
Synonym 2-2-methyl-1h-imidazol-1-yl aniline, 2-2-methyl-imidazol-1-yl-phenylamine, benzenamine,2-2-methyl-1h-imidazol-1-yl, 2-2-methylimidazol-1-yl aniline, 2-2-methylimidazolyl phenylamine, 1-2-amino phenyl-2-methylimidazole, 2-2-methylimidazol-1-yl phenylamine
PubChem CID 13656108
IUPAC Name 2-(2-methylimidazol-1-yl)aniline
SMILES CC1=NC=CN1C1=CC=CC=C1N
Specifications
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 173.22
Percent Purity 97%
Chemical Name or Material 2-(2-Methyl-1H-imidazol-1-yl)aniline