missing translation for 'onlineSavingsMsg'
Learn More

2',3',4'-Trihydroxyacetophenone, 98%

Product Code. 11406947
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11406947 25 g 25g
11496937 5 g 5g
11416947 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11406947 Supplier Thermo Scientific Alfa Aesar Supplier No. A14364.14
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 528-21-2 | C8H8O4 | 168.148 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 528-21-2
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.148
MDL Number MFCD00002193
InChI Key XIROXSOOOAZHLL-UHFFFAOYSA-N
Synonym gallacetophenone, 1-2,3,4-trihydroxyphenyl ethanone, 2',3',4'-trihydroxyacetophenone, galloacetophenone, alizarin yellow c, alizarine yellow c, 2,3,4-trihydroxyacetophenone, ethanone, 1-2,3,4-trihydroxyphenyl, 4-acetylpyrogallol, acetophenone, 2',3',4'-trihydroxy
PubChem CID 10706
IUPAC Name 1-(2,3,4-trihydroxyphenyl)ethanone
SMILES CC(=O)C1=C(C(=C(C=C1)O)O)O

Specifications

Melting Point 169°C to 172°C
Quantity 25 g
Merck Index 14,4342
Formula Weight 168.15
Percent Purity 98%
Chemical Name or Material 2',3',4'-Trihydroxyacetophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.